missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serum Albumin / HSA / ALB Protein
A DNA sequence encoding the Mus musculus (Mouse) Serum Albumin / HSA / ALB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8965-ba-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033784.2 | |
Serum Albumin / HSA / ALB Protein | |
Research Use Only | |
ALB | |
Recombinant Protein | |
Baculovirus | |
EAHKSEIAHR YNDLGEQHFK GLVLIAFSQY LQKCSYDEHA KLVQEVTDFA KTCVADESAA NCDKSLHTLF GDKLCAIPNL RENYGELADC CTKQEPERNE CFLQHKDDNP SLPPFERPEA EAMCTSFKEN PTTFMGHYLH EVARRHPYFY APELLYYAEQ YNEILTQCCA EADKESCLTP KLDGVKEKAL VSSVRQRMKC SSMQKFGERA FKAWAVARLS QTFPNADFAE ITKLATDLTK VNKECCHGDL LECADDRAEL AKYMCENQAT ISSKLQTCCD KPLLKKAHCL SEVEHDTMPA DLPAIAADFV EDQEVCKNYA EAKDVFLGTF LYEYSRRHPD YSVSLLLRLA KKYEATLEKC CAEANPPACY GTVLAEFQPL VEEPKNLVKT NCDLYEKLGE YGFQNAILVR YTQKAPQVST PTLVEAARNL GRVGTKCCTL PEDQRLPCVE DYLSAILNRV CLLHEKTPVS EHVTKCCSGS LVERRPCFSA LTVDETYVPK EFKAETFTFH SDICTLPEKE KQIKKQTALA ELVKHKPKAT AEQLKTVMDD FAQFLDTCCK AADKDTCFST EGPNLVTRCK DALA | |
Greater than 90% as determined by SDS-PAGE. |
11657 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |