missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£384.00 - £1275.00
Specifications
Gene ID (Entrez) | 20423 |
---|---|
Molecular Weight (g/mol) | 19.8 kDa |
Name | Sonic Hedgehog/Shh (C25II), N-Terminus |
Source | E. coli-derived mouse Sonic Hedgehog/Shh protein Cys25-Gly198 (Cys25Ile-Ile),with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
18196180
|
R&D Systems™
464-SH-025 |
25 μg | Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT with BSA as a carrier protein. |
£384.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18157800
|
R&D Systems™
464-SH-025/CF |
25 μg | Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT . |
£384.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18715526
|
R&D Systems™
464-SH-200 |
200 μg | Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT with BSA as a carrier protein. |
£1275.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18776286
|
R&D Systems™
464-SH-200/CF |
200 μg | Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT . |
£1275.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus is derived from E. coli. The Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus has been validated for the following applications: Bioactivity.Specifications
20423 | |
Sonic Hedgehog/Shh (C25II), N-Terminus | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
19.8 kDa | |
E. coli-derived mouse Sonic Hedgehog/Shh protein Cys25-Gly198 (Cys25Ile-Ile),with an N-terminal Met | |
HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title