missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Sphingosine Kinase 1/SPHK1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 6086-SK-010
This item is not returnable.
View return policy
Description
The Recombinant Mouse Sphingosine Kinase 1/SPHK1 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Sphingosine Kinase 1/SPHK1 Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2μm filtered solution in MES, NaCl, Glycerol and DTT. | |
43 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. | |
Unconjugated | |
70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
20698 | |
Sphingosine Kinase 1/SPHK1 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Sphingosine Kinase 1/SPHK1 protein Glu2-Pro382,with an N-terminal Met and 6-His tag | |
Monomer | |
Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction