missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse sTNF RI/TNFRSF1A Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 425-R1-050/CF

276.45 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18146170

  • £357.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse sTNF RI/TNFRSF1A Protein is derived from E. coli. The Recombinant Mouse sTNF RI/TNFRSF1A Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
21 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
21937
TNF RI/TNFRSF1A
E. coli-derived mouse TNF RI/TNFRSF1A protein Ile22-Ala212,with an N-terminal Met
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers