missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TIGIT Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 7267-TG-050
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. When Recombinant Mouse CD155/PVR (Catalog # 6909-CD ) is coated at 2.5 μg/mL (100 μL/well), the concentration of Recombinant Mouse TIGIT Fc Chimera that produces 50% optimal binding response is 10-50 ng/mL.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
NP_001139797 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 50-60 kDa, reducing conditions; M.W. (theoretical): 40.1 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
Reconstitute at 100 μg/mL in PBS. | |
TIGIT |
Bioactivity | |
201633 | |
TIGIT Fc Chimera | |
Mouse myeloma cell line, NS0-derived mouse TIGIT protein Mouse TIGIT (Gly26-Thr143) (N-terminus) Accession # NP_001139797 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Tigit | |
Recombinant | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Mouse TIGIT Fc Chimera Protein > 50ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction