missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TIMP-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 7667-TM-010
283.36 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse TIMP-4 Protein is derived from NS0. The Recombinant Mouse TIMP-4 Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2μm filtered solution in Tris and NaCl. | |
23 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Recombinant |
110595 | |
TIMP-4 | |
Mouse myeloma cell line,NS0-derived mouse TIMP-4 protein Met1-Pro224 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |