missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TLR1 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 1476-TR-050
276.45 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse TLR1 Protein is derived from NS0. The Recombinant Mouse TLR1 Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
64.2 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
21897 | |
TLR1 | |
Mouse myeloma cell line,NS0-derived mouse TLR1 protein Ser25-Asp581,with a C-terminal 10-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |