missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TRANCE/RANK L/TNFSF11 (E. coli-expressed)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£391.00
Specifications
Gene ID (Entrez) | 21943 |
---|---|
Molecular Weight (g/mol) | 19 kDa |
Name | TRANCE/TNFSF11/RANK L |
Quantity | 10 μg |
Source | E. coli-derived mouse TRANCE/TNFSF11/RANK L protein Lys158-Asp316,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18176600
|
R&D Systems™
462-TEC-010 |
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA with BSA as a carrier protein. |
£391.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18359227
|
R&D Systems™
462-TEC-010/CF |
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA. |
£391.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse TRANCE/RANK L/TNFSF11 (E. coli-expressed) is derived from E. coli. The Recombinant Mouse TRANCE/RANK L/TNFSF11 (E. coli-expressed) has been validated for the following applications: Bioactivity.Specifications
21943 | |
TRANCE/TNFSF11/RANK L | |
E. coli-derived mouse TRANCE/TNFSF11/RANK L protein Lys158-Asp316,with an N-terminal Met | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
19 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title