missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse VEGF-B 167 Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

£399.00

Specifications

Gene ID (Entrez) 22339
Molecular Weight (g/mol) 19 kDa (monomer)
Name VEGF-B 167
Quantity 25 μg
Source E. coli-derived mouse VEGF-B protein Pro22-Lys188
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18337497
View Documents
R&D Systems™
2595-VE-025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
£399.00
25µg
Please to purchase this item. Need a web account? Register with us today!
18137020
View Documents Promotion Details
R&D Systems™
2595-VE-025/CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
£399.00
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Mouse VEGF-B 167 Protein is derived from E. coli. The Recombinant Mouse VEGF-B 167 Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

22339
VEGF-B 167
E. coli-derived mouse VEGF-B protein Pro22-Lys188
Disulfide-linked homodimer
Recombinant
19 kDa (monomer)
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Videos
SDS
Documents

Documents

Product Certifications