Learn More
R&D Systems™ Recombinant Mouse Wnt-3a (High Purity) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 1324-WNP-010/CF
This item is not returnable.
View return policy
Description
The Recombinant Mouse Wnt-3a (High Purity) Protein is derived from CHO. The Recombinant Mouse Wnt-3a (High Purity) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS. | |
37.4 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
22416 | |
Wnt-3a (High Purity) | |
Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-3a protein Ser19-Lys352 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |