missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mycoplasma Putative Xaa-Pro aminopeptidase Protein

A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Putative Xaa-Pro aminopeptidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6954-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16096870

  • £458.00 / 50µg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

NP_110158.1
Mycoplasma Putative Xaa-Pro aminopeptidase Protein
Research Use Only
pepP
Recombinant Protein
E. coli
MHNELQQKLA VLHKLLQDNK ADAILIGSDQ NRFWLTGFPS SAGWLVVHKQ RVNLFIDGRY FEAAKTAIDP LVKVELFTTY KQVKALCEQV GVKHLLIEGD YLTFNYQNFI KELCAQYTVI NAQEIRRQKL PSEILAIEKV VEITRKVAVK LKRFIQPGMT ELFIAQWITD QLVKAGGAKN SFDPIVATGK NGANPHHKPS KLKVKSGDFV TCDFGTIYNG YCSDITRTFL VGKKPNNEVL LKAYKKVDEA NMAGINAANT QLTGAEVDKV CRDIIEASEF KDYFVHSTGH GVGLDIHEMP NVSTSYNKLL CENAVITIEP GIYIPSVGGI RIEDMVLVKD HKSVWLSAKI PRAF
Greater than 90% as determined by SDS-PAGE.
877187
50 μg
Not determined.
Full Length
Mycoplasma
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers