missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Bivalirudin Protein
A cDNA sequence encoding the Bivalirudin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11165-5mg
This item is not returnable.
View return policy
Specifications
Bivalirudin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with 0.5 mg Manntiol and sodium hydroxide 50microg pH 5.5. |
5 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |