missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Borrelia BmpA Protein
A cDNA sequence encoding the Borrelia BmpA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11181-20ug
This item is not returnable.
View return policy
Specifications
Borrelia BmpA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Borrelia BmpA (1.3 mg/1ml) is supplied in 20mM HEPES buffer pH 8.0, 6M Urea and 200mM NaCl. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |