missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Borrelia p45 Protein
A cDNA sequence encoding the Borrelia p45 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | Borrelia p45 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15956438
|
enQuireBio™
QP11193-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15946438
|
enQuireBio™
QP11193-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15936438
|
enQuireBio™
QP11193-1mg |
1 mg |
£3965.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Borrelia p45 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
Borrelia p45 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |