missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Phospholipase A2 Protein
A cDNA sequence encoding the Phospholipase A2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13045-500ug
This item is not returnable.
View return policy
Specifications
406141 | |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MQVVLGSLFL LLLSTSHGWQ IRDRIGDNEL EERIIYPGTL WCGHGNKSSG PNELGRFKHT DACCRTHDMC PDVMSAGESK HGLTNTASHT RLSCDCDDKF YDCLKNSADT ISSYFVGKMY FNLIDTKCYK LEHPVTGCGE RTEGRCLHYT VDKSKPKVYQ WFDLRKY | |
Protein is >90% pure as determined by 10% SDS-PAGE. |
Phospholipase A2 Protein | |
Research Use Only | |
Pla2 | |
Other | |
Untagged | |
20mM Tris-HCl, pH 7.2, 1.5M urea and 50% glycerol. |