missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Prelamin-A Protein
A cDNA sequence encoding the Prelamin-A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13135-100ug
This item is not returnable.
View return policy
Specifications
4000 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
metpsqrrat rsgaqasstp lsptritrlq ekedlqelnd rlavyidrvr sletenaglr lriteseevv srevsgikaa yeaelgdark tldsvakera rlqlelskvr eefkelkarn tkkegdliaa qarlkdleal lnskeaalst alsekrtleg elhdlrgqva kleaalgeak kqlqdemlrr vdaenrlqtm keeldfqkni yseelretkr rhetrlveid ngkqrefesr ladalqelra qhedqveqyk kelektysak ldnarqsaer nsnlvgaahe elqqsririd slsaqlsqlq kqlaakeakl rdledslare rdtsrrllae keremaemra rmqqqldeyq elldiklald meihayrkll egeeerlrls psptsqrsrg rasshssqtq gggsvtkkrk lestesrssf sqhartsgrv aveevdeegk fvrlrnksne dqsmgnwqik rqngddpllt yrfppkftlk agqvvtiwaa gagathsppt dlvwkaqntw gcgnslrtal instgeevam rklvrsvtvv eddededgdd llhhhhgshc sssgdpaeyn lrsrtvlcgt cgqpadkasa sgsgaqvggp issgssassv tvtrsyrsvg gsgggsfgdn lvtrsyllgn ssprtqspqn csim | |
Greater than 90% as determined by SDS-PAGE. |
Prelamin-A Protein | |
Research Use Only | |
LMNA | |
Other | |
His | |
The Prelamin-A solution (0.1 mg/ml) contains 10% Glycerol. |