missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other RNASEH2A Protein
A cDNA sequence encoding the RNASEH2A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13326-20ug
This item is not returnable.
View return policy
Specifications
RNASEH2A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDLSELE RDNTGRCRLS SPVPAVCRKE PCVLGVDEAG RGPVLGPMVY AICYCPLPRL ADLEALKVAD SKTLLESERE RLFAKMEDTD FVGWALDVLS PNLISTSMLG RVKYNLNSLS HDTATGLIQY ALDQGVNVTQ VFVDTVGMPE TYQARLQQSF PGIEVTVKAK ADALYPVVSA ASICAKVARD QAVKKWQFVE KLQDLDTDYG SGYPNDPKTK AWLKEHVEPV FGFPQFVRFS WRTAQTILEK EAEDVIWEDS ASENQEGLRK ITSYFLNEGS QARPRSSHRY FLERGLESAT SL | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
The RNASEH2A solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |