missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Terlipressin Protein
A cDNA sequence encoding the Terlipressin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13723-1gram
This item is not returnable.
View return policy
Specifications
Terlipressin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with no additives. |
1 g | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |