missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other TXN2 Yeast Protein
A cDNA sequence encoding the TXN2 Yeast was constructed and used to recombinantly synthesize the protein.
£99.15 - £2645.00
Specifications
Name | TXN2 Yeast Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | TXN2 activity is assayed by measuring the change in absorbance at 650 nm at 25C The specific activity was found to be 3 units/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985518
|
enQuireBio™
QP10903-5ug |
5 μg |
£99.15
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15975518
|
enQuireBio™
QP10903-20ug |
20 μg |
£155.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15965518
|
enQuireBio™
QP10903-1mg |
1 mg |
£2645.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
TXN2 Yeast Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MVTQLKSASE YDSALASGDK LVVVDFFATW CGPCKMIAPM IEKFAEQYSD AAFYKLDVDE VSDVAQKAEV SSMPTLIFYK GGKEVTRVVG ANPAAIKQAI ASNV | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
TXN2 activity is assayed by measuring the change in absorbance at 650 nm at 25C The specific activity was found to be 3 units/mg. | |
Other | |
Untagged | |
Each mg of TRX2 protein contains 20mM phosphate buffer pH 7.4. |