missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa Elastase Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Elastase, was expressed in the hosts and tags indicated. option, ab
Brand: enQuireBio™ QP7029-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_252413.1 | |
P. aeruginosa Elastase Protein | |
Research Use Only | |
lasB | |
Recombinant Protein | |
E. coli | |
AEAGGPGGNQ KIGKYTYGSD YGPLIVNDRC EMDDGNVITV DMNSSTDDSK TTPFRFACPT NTYKQVNGAY SPLNDAHFFG GVVFKLYRDW FGTSPLTHKL YMKVHYGRSV ENAYWDGTAM LFGDGATMFY PLVSLDVAAH EVSHGFTEQN SGLIYRGQSG GMNEAFSDMA GEAAEFYMRG KNDFLIGYDI KKGSGALRYM DQPSRDGRSI DNASQYYNGI DVHHSSGVYN RAFYLLANSP GWDTRKAFEV FVDANRYYWT ATSNYNSGAC GVIRSAQNRN YSAADVTRAF STVGVTCPSA L | |
Greater than 80% as determined by SDS-PAGE. |
880368 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |