missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant P. heparinus Heparinase III Protein
Recombinant Protein
Brand: R&D Systems™ 6145-GH-010
270.40 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to liberate oligosaccharides from heparin. The specific activity is >3,000 pmol/min/μg, as measured under the described conditions.Specifications
AAB18278 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. | |
M.W. (Observed): 66 kDa, reducing conditions; M.W. (theoretical): 74 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
Phep_3797 | |
Recombinant | |
Heparinase III |
Enzyme Activity | |
8254931 | |
Heparinase III | |
E. coli-derived p. heparinus Heparinase III protein Gln25-Pro659, with an N-terminal Met and 6-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.) |