missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine Insulin Protein
A cDNA sequence encoding the Insulin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10733-250mg
Additional Details : Weight : 0.01000kg
Specifications
397415 | |
250 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
>27 units/mg. | |
Porcine | |
Untagged | |
The Insulin Porcine was lyophilized from a concentrated (1 mg/ml) solution with no additives. |
Insulin Protein | |
Research Use Only | |
INS | |
Recombinant Protein | |
Porcine | |
MALWTRLLPL LALLALWAPA PAQAFVNQHL CGSHLVEALY LVCGERGFFY TPKARREAEN PQAGAVELGG GLGGLQALAL EGPPQKRGIV EQCCTSICSL YQLENYCN | |
Greater than 97.0% as determined by SEC-HPLC. |