Learn More
Novex™ Recombinant Protein G
Recombinant Protein G
Brand: Novex™ 101201
244.95 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. It is supplied as a salt-free lyophilized powder.Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:
Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
Specifications
Unconjugated | |
Powder | |
2-8°C |
5 mg | |
Protein G | |
Wet Ice |