Learn More
Novex™ Recombinant Protein G-HRP
Recombinant Protein G-HRP
Brand:  Novex™ 101223
138.32 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10426873
  • £182.00 / 1mL
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This protein G is conjugated with highly purified horseradish peroxidase (RZ > 3.0) and is in enyme conjugate stabilizer containing 0.1% proclin.
Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:
  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG
    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    HRP
    Powder
    2-8°C
    1 mL
    Protein G
    Wet Ice