missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit TNF a Protein
A cDNA sequence encoding the TNF a was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10882-1mg
This item is not returnable.
View return policy
Specifications
100009088 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by the cytolysis of murine L929 cells in the presence of Actinomycin D is less than 0.03ng/ml, corresponding to a Specific Activity of 30,000,000 IU/mg. | |
Rabbit | |
Untagged | |
TNF-alpha Rabbit was lyophilized after extensive dialysis against 20mM PBS, pH 7.4, 300mM NaCl. |
TNF a Protein | |
Research Use Only | |
TNF | |
Recombinant Protein | |
E. coli | |
MSTESMIRDV ELAEGPLPKK AGGPQGSKRC LCLSLFSFLL VAGATTLFCL LHFRVIGPQE EEQSPNNLHL VNPVAQMVTL RSASRALSDK PLAHVVANPQ VEGQLQWLSQ RANALLANGM KLTDNQLVVP ADGLYLIYSQ VLFSGQGCRS YVLLTHTVSR FAVSYPNKVN LLSAIKSPCH RETPEEAEPM AWYEPIYLGG VFQLEKGDRL STEVNQPEYL DLAESGQVYF GIIAL | |
Greater than 95% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |