missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat ANPEP / APN / CD13 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) ANPEP / APN / CD13, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5648-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_112274.1 | |
ANPEP / APN / CD13 Protein | |
Research Use Only | |
Anpep | |
Recombinant Protein | |
E. coli | |
YAQEKNRNAE NSAIAPTLPG STSATTSTTN PAIDESKPWN QYRLPKTLIP DSYQVTLRPY LTPNEQGLYI FKGSSTVRFT CNETTNVIII HSKKLNYTNK GNHRVALRAL GDTPAPNIDT TELVERTEYL VVHLQGSLVK GHQYEMDSEF QGELADDLAG FYRSEYMEGG NKKVVATTQM QAADARKSFP CFDEPAMKAS FNITLIHPNN LTALSNMLPK DSRTLQEDPS WNVTEFHPTP KMSTYLLAYI VSEFKYVEAV SPNRVQIRIW ARPSAIDEGH GDYALQVTGP ILNFFAQHYN TAYPLEKSDQ IALPDFNAGA MENWGLVTYR ESALVFDPQS SSISNKERVV TVIAHELAHQ WFGNLVTVDW WNDLWLNEGF ASYVEFLGAD YAEPTWNLKD LIVLNDVYRV MAVDALASSH PLSSPANEVN TPAQISELFD SITY | |
Greater than 90% as determined by SDS-PAGE. |
81641 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |