missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat beta-NGF / Beta-NGF Protein
A DNA sequence encoding the Rattus norvegicus (Rat) beta-NGF / Beta-NGF Protein, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_001263984.1 |
---|---|
Gene ID (Entrez) | 310738 |
Name | beta-NGF / Beta-NGF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16078061
|
enQuireBio™
QP9305-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018071
|
enQuireBio™
QP9305-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088061
|
enQuireBio™
QP9305-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008071
|
enQuireBio™
QP9305-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028071
|
enQuireBio™
QP9305-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098061
|
enQuireBio™
QP9305-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001263984.1 | |
beta-NGF / Beta-NGF Protein | |
Not determined. | |
Full. Length. of. Mature. Protein. | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
310738 | |
Research Use Only | |
Ngf | |
Recombinant Protein | |
Yeast | |
SSTHPVFHMG EFSVCDSVSV WVGDKTTATD IKGKEVTVLG EVNINNSVFK QYFFETKCRA PNPVESGCRG IDSKHWNSYC TTTHTFVKAL TTDDKQAAWR FIRIDTACVC VLSRKAARRG | |
Greater than 90% as determined by SDS-PAGE. |