missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Carbonic anhydrase 1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Carbonic anhydrase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5741-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001101130.1 | |
Carbonic anhydrase 1 Protein | |
Research Use Only | |
Ca1 | |
Recombinant Protein | |
E. coli | |
ASADWGYDSK NGPDQWSKLY PIANGNNQSP IDIKTSEAKH DSSLKPVSVS YNPATAKEIV NVGHSFHVVF DDSSNQSVLK GGPLADSYRL TQFHFHWGNS NDHGSEHTVD GAKYSGELHL VHWNSAKYSS AAEAISKADG LAIIGVLMKV GPANPNLQKV LDALSSVKTK GKRAPFTNFD PSSLLPSSLD YWTYFGSLTH PPLHESVTWV ICKESISLSP EQLAQLRGLL SSAEGEPAVP VLSNHRPPQP LKGRTVRASF | |
Greater than 90% as determined by SDS-PAGE. |
310218 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |