missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL2 / MCP-1 / MCP1 Protein
Recombinant Protein
Brand: enQuireBio™ QP5509-2ug
This item is not returnable.
View return policy
Specifications
NP_113718.1 | |
CCL2 / MCP-1 / MCP1 Protein | |
Research Use Only | |
CCL2 | |
Recombinant Protein | |
E. coli | |
QPDAVNAPLT CCYSFTGKMI PMSRLENYKR ITSSRCPKEA VVFVTKLKRE ICADPNKEWV QKYIRKLDQN QVRSETTVFY KIASTLRTSA PLNVNLTHKS EANASTLFST TTSSTSVEVT SMTEN | |
Greater than 95% as determined by SDS-PAGE. |
24770 | |
2 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat CCL2 / MCP-1 / MCP1 induces THP-1 chemotaxis (significanty different from control at Rat CCL2 / MCP-1 / MCP1 concentrations greater than 10 ng/ml). | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |