missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Cyclophilin F Protein
A cDNA sequence encoding the Cyclophilin F was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10573-2ug
This item is not returnable.
View return policy
Specifications
282819 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 1,300 nmol/min/mg, and is defined as the amount of enzyme that cleaves 1nmol of suc-AAFP-PNA per minute at 37C in Tris–HCl pH 8.0 using chymotrypsin. | |
Rat | |
His | |
Cyclophilin F protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 10% glycerol and 1mM DTT. |
Cyclophilin F Protein | |
Research Use Only | |
Ppif | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSCSDGGAR GANSSSQNPL VYLDVGADGQ PLGRVVLELK ADVVPKTAEN FRALCTGEKG FGYKGSTFHR VIPAFMCQAG DFTNHNGTGG KSIYGSRFPD ENFTLKHVGP GVLSMANAGP NTNGSQFFIC TIKTDWLDGK HVVFGHVKEG MDVVKKIESF GSKSGKTSKK IVITDCGQLS | |
Greater than 90.0% as determined by SDS-PAGE. |