Learn More
R&D Systems™ Recombinant Rat Dkk-1 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 4010-DK-010/CF
 View more versions of this product
Product Code. 18394827
  • £399.56 / 10µg
Explore more special offers
Description
Description
The Recombinant Rat Dkk-1 Protein is derived from NS0.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
Dkk-1
Mouse myeloma cell line,NS0-derived rat Dkk-1 protein Ser23-His270,with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
27.6 kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Product Suggestions