missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat FGF basic/FGF2/bFGF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 3339-FB-025
183.15 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Rat FGF basic/FGF2 Protein is derived from E. coli. The Recombinant Rat FGF basic/FGF2 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein. | |
FGF basic | |
E. coli-derived rat FGF basic/FGF2/bFGF protein Ala11-Ser154 | |
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin | |
Recombinant |
16.2 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |