missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Glypican-3 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Glypican-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9158-ye-10ug
This item is not returnable.
View return policy
Specifications
NP_036906.1 | |
Glypican-3 Protein | |
Research Use Only | |
GPC3 | |
Recombinant Protein | |
Yeast | |
QPPPPPDATC HQVRSFFQRL QPGLKWVPET PVPGSDLQVC LPKGPTCCSR KMEEKYQLTA RLNMEQLLQS ASMELKFLII QNAAVFQEAF EIVVRHAKNY TNAMFKNNYP SLTPQAFEFV GEFFTDVSLY ILGSDINVDD MVNELFDSLF PVIYTQMMNP GLPESVLDIN ECLRGARRDL KVFGSFPKLI MTQVSKSLQV TRIFLQALNL GIEVINTTDH LKFSKDCGRM LTRMWYCSYC QGLMMVKPCG GYCNVVMQGC MAGVVEIDKY WREYILSLEE LVNGMYRIYD MENVLLGLFS TIHDSIQYVQ KNGGKLTTTI GKLCAHSQQR QYRSAYYPED LFIDKKVLKV ARVEHEETLS SRRRELIQKL KSFISFYSAL PGYICSHSPV AENDTLCWNG QELVERYSQK AARNGMKNQF NLHELKMKGP EPVVSQIIDK LKHINQLLRT MSVPKGKVVD KSLDEEGLES GDCGDDEDEC IGSSGDGMMK VKNQLRFLAE LAYDLDVDDA PGNKQHGNQK DNEITTSHSV GNMPSP | |
Greater than 90% as determined by SDS-PAGE. |
25236 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |