missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat MAPT / Tau Protein

A DNA sequence encoding the Rattus norvegicus (Rat) MAPT / Tau, was expressed in the hosts and tags indicated.

£223.39 - £2410.48

Specifications

Name MAPT / Tau Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Mapt
Protein Length Full Length
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16042630
View Documents Promotion Details
enQuireBio™
QP6345-YE-100UG
100 μg
£560.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-09-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16082630
View Documents Promotion Details
enQuireBio™
QP6345-YE-500UG
500 μg
£1600.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 05-09-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16052630
View Documents Promotion Details
enQuireBio™
QP6345-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16072630
View Documents Promotion Details
enQuireBio™
QP6345-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16092630
View Documents Promotion Details
enQuireBio™
QP6345-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16062630
View Documents Promotion Details
enQuireBio™
QP6345-YE-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

MAPT / Tau Protein
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Research Use Only
Mapt
Recombinant Protein
Yeast
AEPRQEFDTM EDQAGDYTML QDQEGDMDHG LKESPPQPPA DDGSEEPGSE TSDAKSTPTA EDVTAPLVEE RAPDKQATAQ SHTEIPEGTT AEEAGIGDTP NMEDQAAGHV TQEPQKVEIF SQSLLVEPGR REGQAPDSGI SDWTHQQVPS MSGAPLPPQG LREATHQPLG TRPEDVERSH PASELLWQES PQKEAWGKDR LGSEEEVDED ITMDESSQES PPSQASLAPG TATPQARSVS ASGVSGETTS IPGFPAEGSI PLPADFFSKV SAETQASPPE GPGTGPSEEG HEAAPEFTFH VEIKASAPKE QDLEGATVVG APAEEQKARG PSVGKGTKEA SLLEPTDKQP AAGLPGRPVS RVPQLKARVA GVSKDRTGND EKKAKTSTPS CAKTPSNRPC LSPTRPTPGS SDPLIKPSSP AVCPEPATSP KYVSSVTPRN GSPGTKQMKL KGADGKTGAK IATPRGAATP GQKGTSNATR IPAKTTPSPK TPPGSGEPPK SGERSGYSSP GSPGTPGSRS RTPSLPTPPT REPKKVAVVR TPPKSPSASK SRLQTAPVPM PDLKNVRSKI GSTENLKHQP GGGKVQIINK KLDLSNVQSK CGSKDNIKHV PGGGSVHIVY KPVDLSKVTS KCGSLGNIHH KPGGGQVEVK SEKLDFKDRV QSKIGSLDNI THVPGGGNKK IETHKLTFRE NAKAKTDHGA EIVYKSPVVS GDTSPRHLSN VSSTGSIDMV DSPQLATLAD EVSASLAKQG L
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications