missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Minpp1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Minpp1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6360-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_062136.1 | |
Minpp1 Protein | |
Research Use Only | |
Minpp1 | |
Recombinant Protein | |
E. coli | |
SLPGRGDPVA SVLSPYFGTK TRYEDVNPWL LGDPVAPRRD PELLAGTCTP VQLVALIRHG TRYPTTKQIR KLRQLQGLLQ TRESVDGGSR VAAALDQWPL WYDDWMDGQL VEKGRQDMRQ LALRLAALFP DLFCRENYGR LRLITSSKHR CVDSSAAFLQ GLWQHYHPGL PPPDVSDMEC DPPRVNDKLM RFFDHCEKFL TEVERNATAL YHVEAFKTGP EMQTVLKKVA ATLQVPVNNL NADLIQVAFF TCSFDLAIQG VHSPWCDVFD VDDAKVLEYL NDLKQYWKRS YGYAINSRSS CNLFQDIFLH LDKAVEQKQR SQPVSSSVIL QFGHAETLLP LLSLMGYFKD KEPLTAYNFE EQVHREFRSG HIVPYASNLI FVLYHCEDAQ TPQEKFQIQM LLNEKVLPLA HSQKTVALYE DLKNHYQDIL QSCQTSKECN LPKVNITSDE L | |
Greater than 90% as determined by SDS-PAGE. |
29688 | |
1 mg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |