missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Muscarinic acetylcholine receptor M1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Muscarinic acetylcholine receptor M1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5831-ye-200ug
This item is not returnable.
View return policy
Specifications
NP_542951.1 | |
Muscarinic acetylcholine receptor M1 Protein | |
Research Use Only | |
Chrm1 | |
Recombinant Protein | |
Yeast | |
RIYRETENRA RELAALQGSE TPGKGGGSSS SSERSQPGAE GSPESPPGRC CRCCRAPRLL QAYSWKEEEE EDEGSMESLT SSEGEEPGSE VVIKMPMVDS EAQAPTKQPP KSSPNTVKRP TKKGRDRGGK GQKPRGKEQL AKRKTFSLVK EKKAART | |
Greater than 90% as determined by SDS-PAGE. |
25229 | |
200 μg | |
Not determined. | |
Cytoplasmic Domain | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |