missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Osteocrin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Osteocrin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6455-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_997495.1 | |
Osteocrin Protein | |
Research Use Only | |
Ostn | |
Recombinant Protein | |
E. coli | |
SFSGFGSPLD RLSAGSVEHR GKQRRVVDHS KKRFGIPMDR IGRNRLSSSR | |
Greater than 90% as determined by SDS-PAGE. |
360730 | |
100 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |