missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Osteocrin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Osteocrin, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_997495.1 |
---|---|
Gene ID (Entrez) | 360730 |
Name | Osteocrin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16073420
|
enQuireBio™
QP6455-ec-10ug |
10 μg | |||||||
16013430
|
enQuireBio™
QP6455-ec-50ug |
50 μg | |||||||
16063420
|
enQuireBio™
QP6455-ec-100ug |
100 μg |
£560.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16093420
|
enQuireBio™
QP6455-ec-200ug |
200 μg | |||||||
16003430
|
enQuireBio™
QP6455-ec-500ug |
500 μg | |||||||
16083420
|
enQuireBio™
QP6455-ec-1mg |
1 mg |
£2480.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_997495.1 | |
Osteocrin Protein | |
Not determined. | |
Full Length | |
Rat | |
Tris-based buffer, 50% glycerol |
360730 | |
Research Use Only | |
Ostn | |
Recombinant Protein | |
SFSGFGSPLD RLSAGSVEHR GKQRRVVDHS KKRFGIPMDR IGRNRLSSSR |