missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Suppressor of cytokine signaling 3 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Suppressor of cytokine signaling 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6713-ye-200ug
This item is not returnable.
View return policy
Specifications
NP_446017.1 | |
Suppressor of cytokine signaling 3 Protein | |
Research Use Only | |
Socs3 | |
Recombinant Protein | |
Yeast | |
MVTHSKFPAA GMSRPLDTSL RLKTFSSKSE YQLVVNAVRK LQESGFYWSA VTGGEANLLL SAEPAGTFLI RDSSDQRHFF TLSVETQSGT KNLRIQCEGG SFSLQSDPRS TQPVPRFDCV LKLVHHYMPP PGAPSFSLPP TEPSFEVQEQ PPAQALPGGT PKRAYYIYSG GEKIPLVLSR PLSSNVATLQ HLCRKTVNGH LDSYEKVTQL PGPIREFLDQ YDAPL | |
Greater than 90% as determined by SDS-PAGE. |
89829 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |