missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Thyroglobulin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Thyroglobulin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6776-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
Thyroglobulin Protein | |
Research Use Only | |
Tg | |
Recombinant Protein | |
E. coli | |
NIFEYQVDAQ PLRPCELQRE KAFLKQDEYV PQCSEDGSFQ TVQCQNDGQS CWCVDSDGTE VPGSRQLGRP TACLSFCQLH KQRILLSSYI NSTDALYLPQ CQDSGNYAPV QCDLQQVQCW CVDTEGMEVY GTRQQGRPTR CPRSCEIRSR RLLHGVGDKS PPQCDADGEF MPVQCKFVNT TDMMIFDLIH NYNRFPDAFV TFSAFRNRFP EVSGYCYCAD SQGRELAETG LELLLDEIYD TIFAGLDQAS TFTQSTMYRI LQRRFLAIQL VISGRFRCPT | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |