missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque IL 3 Protein
A cDNA sequence encoding the IL 3 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | IL 3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The ED50 as determined by the dose-dependant stimulation of the proliferation of human TF-1 cells is less than 0.1ng/ml, corresponding to a Specific Activity of 10,000,000IU/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944938
|
enQuireBio™
QP10711-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15924938
|
enQuireBio™
QP10711-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15934938
|
enQuireBio™
QP10711-1mg |
1 mg |
£2975.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
IL 3 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
APMTQTTSLK TSWAKCSNMI DEIITHLNQP PLPSPDFNNL NEEDQTILVE KNLRRSNLEA FSKAVKSLQN ASAIESILKN LPPCLPMATA APTRPPIRIT NGDRNDFRRK LKFYLKTLEN EQAQ | |
Greater than 97.0% as determined by SDS-PAGE. |
Research Use Only | |
The ED50 as determined by the dose-dependant stimulation of the proliferation of human TF-1 cells is less than 0.1ng/ml, corresponding to a Specific Activity of 10,000,000IU/mg. | |
Rhesus Macaque | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |