missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Clumping factor A Protein
A DNA sequence encoding the Staphylococcus aureus (strain COL) Clumping factor A, was expressed in the hosts and tags indicated.
£321.05 - £2965.00
Specifications
Accession Number | WP_001056223.1 |
---|---|
Name | S. aureus Clumping factor A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | clfA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16040091
|
enQuireBio™
QP7668-ye-10ug |
10 μg | |||||||
16080091
|
enQuireBio™
QP7668-ye-50ug |
50 μg | |||||||
16030091
|
enQuireBio™
QP7668-ye-100ug |
100 μg |
£800.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16060091
|
enQuireBio™
QP7668-ye-200ug |
200 μg | |||||||
16070091
|
enQuireBio™
QP7668-ye-500ug |
500 μg | |||||||
16050091
|
enQuireBio™
QP7668-ye-1mg |
1 mg |
£2965.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
WP_001056223.1 | |
Research Use Only | |
clfA | |
Recombinant Protein | |
Yeast | |
GTDITNQLTN VTVGIDSGTT VYPHQAGYVK LNYGFSVPNS AVKGDTFKIT VPKELNLNGV TSTAKVPPIM AGDQVLANGV IDSDGNVIYT FTDYVNTKDD VKATLTMPAY IDPENVKKTG NVTLATGIGS TTANKTVLVD YEKYGKFYNL SIKGTIDQID KTNNTYRQTI YVNPSGDNVI APVLTGNLKP NTDSNALIDQ QNTSIKVYKV DNAADLSESY FVNPENFEDV TNSVNITFPN PNQYKVEFNT PDDQITTPYI VVVNGHIDPN SKGDLALRST LYGYNSNIIW RSMSWDNEVA FNNGSGSGDG IDKPVVPEQP DEPGEIEPIP E | |
Greater than 95% as determined by SDS-PAGE. |
S. aureus Clumping factor A Protein | |
Not determined. | |
Partial (see sequence information for more details). Protein | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |