missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Enterotoxin type D Protein

A DNA sequence encoding the Staphylococcus aureus Enterotoxin type D, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7099-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037370

  • £780.00 / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

S. aureus Enterotoxin type D Protein
Research Use Only
entD
Recombinant Protein
E. coli
NENIDSVKEK ELHKKSELSS TALNNMKHSY ADKNPIIGEN KSTGDQFLEN TLLYKKFFTD LINFEDLLIN FNSKEMAQHF KSKNVDVYPI RYSINCYGGE IDRTACTYGG VTPHEGNKLK ERKKIPINLW INGVQKEVSL DKVQTDKKNV TVQELDAQAR RYLQKDLKLY NNDTLGGKIQ RGKIEFDSSD GSKVSYDLFD VKGDFPEKQL RIYSDNKTLS TEHLHIDIYL YEK
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers