missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type D Protein
A DNA sequence encoding the Staphylococcus aureus Enterotoxin type D, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7099-500ug
Additional Details : Weight : 0.01000kg
Specifications
S. aureus Enterotoxin type D Protein | |
Research Use Only | |
entD | |
Recombinant Protein | |
E. coli | |
NENIDSVKEK ELHKKSELSS TALNNMKHSY ADKNPIIGEN KSTGDQFLEN TLLYKKFFTD LINFEDLLIN FNSKEMAQHF KSKNVDVYPI RYSINCYGGE IDRTACTYGG VTPHEGNKLK ERKKIPINLW INGVQKEVSL DKVQTDKKNV TVQELDAQAR RYLQKDLKLY NNDTLGGKIQ RGKIEFDSSD GSKVSYDLFD VKGDFPEKQL RIYSDNKTLS TEHLHIDIYL YEK | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |