missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Staphylococcal complement inhibitor Protein
A DNA sequence encoding the Staphylococcus aureus (strain N315) Staphylococcal complement inhibitor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8018-ec-10ug
This item is not returnable.
View return policy
Specifications
WP_000702262.1 | |
10 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus Staphylococcal complement inhibitor Protein | |
Research Use Only | |
scn | |
Recombinant Protein | |
E. coli | |
STSLPTSNEY QNEKLANELK SLLDELNVNE LATGSLNTYY KRTIKISGLK AMYALKSKDF KKMSEAKYQL QKIYNEIDEA LKSKY | |
Greater than 90% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction