missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae DDP1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) DDP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8030-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_014806.1 | |
S. cerevisiae DDP1 Protein | |
Research Use Only | |
DDP1 | |
Recombinant Protein | |
E. coli | |
GKTADNHGPV RSETAREGRE NQVYSPVTGA RLVAGCICLT PDKKQVLMIT SSAHKKRWIV PKGGVEKDEP NYETTAQRET WEEAGCIGKI VANLGTVEDM RPPKDWNKDI KQFENSRKDS EVAKHPPRTE FHFYELEIEN LLDKFPECHK RHRKLYSYTE AKQNLIDAKR PELLEALNRS AIIKDDK | |
Greater than 90% as determined by SDS-PAGE. |
854334 | |
10 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |