missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Salmon Calcitonin Protein
A cDNA sequence encoding the Calcitonin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11261-5mg
This item is not returnable.
View return policy
Specifications
Salmon Calcitonin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Calcitonin Acetate (Salmon) has an amino acid sequence of: Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2 | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
5 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Salmon | |
Untagged | |
The calcitonin peptide was lyophilized with no additives. |