missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Salmonella Typhi S. Typhi H Protein
A cDNA sequence encoding the S. Typhi H was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13382-500ug
This item is not returnable.
View return policy
Specifications
Salmonella Typhi S. Typhi H Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile Filtered solution containing PBS and 25mM arginine. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Salmonella Typhi | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |