missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Salmonella Typhi S. Typhi OMP 52kDa Protein

A cDNA sequence encoding the S. Typhi OMP 52kDa was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13385-500ug

 View more versions of this product

Product Code. 15963399

  • £439.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Salmonella Typhi S. Typhi OMP 52kDa Protein
Research Use Only
Recombinant Protein
E. coli
Lyophilized from 1 mg/ml in 20mM sodium carbonate pH 10.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Salmonella Typhi
His
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers