missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Salmonella Typhi S. Typhi OMP 52kDa Protein
A cDNA sequence encoding the S. Typhi OMP 52kDa was constructed and used to recombinantly synthesize the protein.
£155.00 - £660.00
Specifications
Name | Salmonella Typhi S. Typhi OMP 52kDa Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Salmonella Typhi |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943399
|
enQuireBio™
QP13385-100ug |
100 μg |
£155.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15963399
|
enQuireBio™
QP13385-500ug |
500 μg |
£439.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15953399
|
enQuireBio™
QP13385-1mg |
1 mg |
£660.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Salmonella Typhi S. Typhi OMP 52kDa Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Salmonella Typhi | |
His | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized from 1 mg/ml in 20mM sodium carbonate pH 10. |